Learn More
Micro Particles Based on Melamine Resin, Carboxylate-Modified, FITC-marked, Analytical Standard, Size: 1 m, MilliporeSigma™ Supelco™
Fluorescent marker for macrostructures, e.g. microtubules; Aldehyde modification for coupling with amines
Supplier: MilliporeSigma Supelco 862965MLF
Description
Specifications
2.5% (solids) | |
5 mL |