Learn More
Fisherbrand™ Micro Stir Bars Products for Science Education
Micro Color-Coded Stirring Bar For stirring test tubes, micro-beakers, etc.
 View more versions of this product
Catalog No. S29171
Add to cart
Description
Description
  • Octagonal shape
  • Resists corrosion, heat, and cold
  • Available in blue, red or yellow
  • PTFE
Specifications
Specifications
2 mm
PTFE (Polytetrafluoroethylene)
Octagonal
2.2 mm
Red
0.07 in.
0.27 in.
7 mm
12
0.09 in.
Yes
Stir Bar Micro Red 7x2 mm
0.27 x 0.09 in. (7 x 2.2 mm)