Learn More
Applied Biosystems™ MicroAmp™ 96-Well Reaction Tube/Tray/Retainer Set, 0.2 mL
Products 2
Catalog Number Mfr. No. Description Price Qty Availability  
Catalog Number Mfr. No. Description Price Qty Availability  
40-308-6 Non-distribution product as customer accommodation.
View Documents
Applied Biosystems™
403086
N/A 100 assemblies N/A
40-308-3 Non-distribution product as customer accommodation.
View Documents
Applied Biosystems™
403083
N/A 20 assemblies N/A
Description
Description
Optimally designed for precise PCR on Applied Biosystems 0.2mL thermal cyclers.
  • Economical 100-unit bulk packs
  • Nonsterile and autoclavable
Greater Sample Uniformity
  • Use with all instruments that have heated sample covers to eliminate condensation and the need for a mineral oil overlay
  • Proprietary MicroAmp Reaction Tubes are essential for high-performance PCR and ensure the greatest temperature and amplification uniformity for samples
See user manual or package insert for limited label license and trademark information.
PCR, PCR and Real-Time PCR, Real Time PCR (qPCR)