Learn More
NuGen Technologies IncSupplier Diversity Partner MICROARRAY FRAG&LABEL 12  DFS Item
Manufacturer:  NuGen Technologies IncSupplier Diversity Partner 420012
The Encore Biotin Module is used for fragmentation and labeling of cDNA for analysis on Affymetrix Arrays. This product provides a rapid, simple, and automatable approach to fragmentation and labeling of up to five ug of cDNA
Catalog No. 50-648-072