Learn More
NuGen Technologies IncSupplier Diversity Partner MICROARRAY PICO 12  DFS Item
Manufacturer:  NuGen Technologies IncSupplier Diversity Partner 330212
The Ovation Pico WTA System V2 is a whole transcriptome amplification solution for preparing amplified cDNA from an input range of 500 pg to 50 ng total RNA, with yields of over 6 ug cDNA. Recommended for Affymetrix arrays
Catalog No. 50-648-065