Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Microbe Scavenger Hunt
Explore the microworld in your environment.
Specifications
For Use With (Application) | Hunt for various kinds of microorganisms |
---|
Includes:
- Petri dishes with “ready-to-pour” media
- “Microlife traps” for soil and water
- Inoculating loops
- Microscope slide and cover slips
- Stain set
- Pipets
- Dichotomous key to molds, bacteria and protists
- Guide to appropriate viewing techniques
- Comprehensive teacher and student guides
Requires: Compound microscope
Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
S32006
![]() |
Neo/Sci Corporation
201073 |
Lab Investigation Kit |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
Description
Hunt for various kinds of microorganisms including bacteria, fungi and protists in water, air, soil and even food, employing a ready-to-use growth medium.
- Utilizes a specially formulated gel, which requires no autoclaving or special equipment
- Makes it easy for students to design experiments that explore the subvisible world around them by collecting and identifying a wide variety of microbes
Order Info
Class size: 40 students.
Specifications
Hunt for various kinds of microorganisms |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title