Learn More
Bon Opus Biosciences MICROBIAL DNA ENRICHKIT 24RXNS
SDP
Manufacturer:  Bon Opus Biosciences DF253ENRICH KIT CS 1
Encompass_Preferred
Microbial DNA Enrichment Kit (24 rxns RUO) with Devin® filter for fast host DNA depletion. Compatible with downstream applications including NGS on all platforms, qPCR and end point PCR
Catalog No. NC2058321