Learn More
Microbiologics™ BD Phoenix NMIC/ID QC Set
Microorganism sets for culture purposes and QC applications
Manufacturer:  Microbiologics™ 5147L
 View more versions of this product
Catalog No. 23-017-074
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Multiple
Culture purpose and QC application
Escherichia coli ATCC 25922, Pseudomonas aeruginosa ATCC 27853, Escherichia coli ATCC 35218, Klebsiella pneumoniae subspecies pneumoniae ATCC 700603
LYFO DISK
Multiple
CE, online certificate of analysis
Lyophilized Pellet
Lyfo disk™
6/Pk.
2° to 8°C
Safety and Handling
ShelfLife : 6 to 24 Months