Microbiologics™ Big 6 STEC QC Set

Microorganism sets for culture purposes and QC applications

Manufacturer: Microbiologics™ 5218L

 View more versions of this product

Catalog No. 23-017-153

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Specifications

    For Use With (Application) Culture purpose and QC application
    Format Lyophilized Pellet
    Includes Escherichia coli serotype O26:H11 CDC 03-3014, Escherichia coli serotype O45:H2 CDC 00-3039, Escherichia coli serotype O103:H11 CDC 06-3008, Escherichia coli serotype O111:H8 CDC 2010C-3114, Escherichia coli serotype O121:H19 CDC 02-3211, Escherichia coli serotype O145:NM CDC 99-3311
    Media Type Lyfo disk™
    Product Type Big 6 STEC QC set
    Quantity 6 Pellets
    Shelf Life 6 to 24 Months
    Storage Requirements 2° to 8°C
    Certifications/Compliance CE, online certificate of analysis

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit