Learn More
Microbiologics™ Big 6 STEC QC Set
Microorganism sets for culture purposes and QC applications
Manufacturer:  Microbiologics™ 5218K
 View more versions of this product
Catalog No. 23-017-151
Due to product restrictions, please to purchase or view availability for this product.
Specifications Description & Specifications
Specifications
CE, online certificate of analysis
Lyophilized Pellet
Kwik-stik™
6U
Culture purpose and QC application
Escherichia coli serotype O26:H11 CDC 03-3014, Escherichia coli serotype O45:H2 CDC 00-3039, Escherichia coli serotype O103:H11 CDC 06-3008, Escherichia coli serotype O111:H8 CDC 2010C-3114, Escherichia coli serotype O121:H19 CDC 02-3211, Escherichia coli serotype O145:NM CDC 99-3311
Big 6 STEC QC set
2° to 8°C
Safety and Handling
ShelfLife : 6 to 24 Months