Learn More
Microbiologics™ Big 7 STEC QC Set
Microorganism sets for culture purposes and QC applications
Manufacturer:  Microbiologics™ 5219L
 View more versions of this product
Catalog No. 23-017-156
Due to product restrictions, please to purchase or view availability for this product.
Disclaimers:
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. MicroBioLogics Inc. is licensed to use these trademarks and to sell products derived from ATCC cultures.
Specifications Description & Specifications
Specifications
CE, online certificate of analysis
Lyophilized Pellet
Lyfo disk™
6 Pellets
Culture purpose and QC application
Escherichia coli serotype O26:H11 CDC 03-3014, Escherichia coli serotype O45:H2 CDC 00-3039, Escherichia coli serotype O103:H11 CDC 06-3008, Escherichia coli serotype O111:H8 CDC 2010C-3114, Escherichia coli serotype O121:H19 CDC 02-3211, Escherichia coli serotype O145:NM CDC 99-3311, Escherichia coli serotype O157:H7 ATCC 35150
Big 7 STEC QC set
2° to 8°C
Safety and Handling
ShelfLife : 6 to 24 Months