Learn More
Microbiologics™ BIOLOG FF Microplate QC Set
Microorganism sets for culture purposes and QC applications
Manufacturer:  Microbiologics™ 5171P
 View more versions of this product
Catalog No. 23-004-040
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Multiple
Culture purpose and QC application
Penicillium chrysogenum ATCC 10106, Aspergillus niger ATCC 16888, Geotrichum candidum ATCC 34614
Biolog FF microplate QC set
Multiple
CE, online certificate of analysis
Lyophilized Pellet
Kwik-stik™
2U
2° to 8°C
Safety and Handling
ShelfLife : 6 to 24 Months