Learn More
Microbiologics™ CLSI M100 Modified Hodge Test QC Set
Microorganism sets for culture purposes and QC applications
Manufacturer:  Microbiologics™ 5205P
 View more versions of this product
Catalog No. 23-001-937
Due to product restrictions, please to purchase or view availability for this product.
Specifications Description & Specifications
Specifications
CE, online certificate of analysis
Lyophilized Pellet
Kwik-stik™
2U
Culture purpose and QC application
Klebsiella pneumoniae ATCC BAA-1705, Klebsiella pneumoniae ATCC BAA-1706
CLSI M100 MODIFIED HODGE test QC set
2° to 8°C
Safety and Handling
ShelfLife : 6 to 24 Months