Learn More
Microbiologics™ CLSI (NCCLS) M22; Campylobacter Agar QC Set
Microorganism sets for culture purposes and QC applications
Manufacturer:  Microbiologics™ 5026K
 View more versions of this product
Catalog No. 23-017-027
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
25922?*,33291?*
Culture purpose and QC application
Escherichia coli ATCC 25922, Campylobacter jejuni subspecies jejuni ATCC 33291
KWIK-STIK
Escherichia coli, Campylobacter jejuni subsp. jejuni
CE, online certificate of analysis
Lyophilized Pellet
Kwik-stik™
6/Pk.
2° to 8°C
Safety and Handling
ShelfLife : 6 to 24 Months