Learn More
Microbiologics™ CLSI (NCCLS) M22; Chocolate Agar QC Set
Microorganism sets for culture purposes and QC applications
Manufacturer:  Microbiologics™ 5027K
 View more versions of this product
Catalog No. 23-017-029
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Multiple
Culture purpose and QC application
Neisseria gonorrhoeae ATCC 43069, Haemophilus influenzae (type b) ATCC 10211
KWIK-STIK
Multiple
CE, online certificate of analysis
Lyophilized Pellet
Kwik-stik™
6/Pk.
2° to 8°C
Safety and Handling
ShelfLife : 6 to 24 Months