Learn More
Microbiologics™ DuPont™ Qualicon RiboPrinter™ QC Set
Microorganism sets for culture purposes and QC applications
Manufacturer:  Microbiologics™ 5183L
 View more versions of this product
Catalog No. 23-017-090
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Multiple
Culture purpose and QC application
Enterobacter aerogenes ATCC 13048, Staphylococcus epidermidis ATCC 14990, Bacillus pumilus ATCC 700814, Citrobacter braakii ATCC 51113
LYFO DISK
Multiple
CE, online certificate of analysis
Lyophilized Pellet
Lyfo disk™
6/Pk.
2° to 8°C
Safety and Handling
ShelfLife : 6 to 24 Months