Learn More
Microbiologics™ DuPont™ Qualicon RiboPrinter™ QC Set
Microorganism sets for culture purposes and QC applications
Manufacturer:  Microbiologics™ 5183P
 View more versions of this product
Catalog No. 23-004-806
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Multiple
Culture purpose and QC application
Enterobacter aerogenes ATCC 13048, Staphylococcus epidermidis ATCC 14990, Bacillus pumilus ATCC 700814, Citrobacter braakii ATCC 51113
KWIK-STIK
Multiple
CE, online certificate of analysis
Lyophilized Pellet
Kwik-stik™
2/Pk.
2° to 8°C
Safety and Handling
ShelfLife : 6 to 24 Months