Learn More
Microbiologics™ Epower™ Clostridium sporogenes ATCC™ 19404™†
For performing quality control functions including microbial detection, equipment calibration, method validation, and bioburden determination
Specifications
Quantity 10/Pk.
View More Specs
Products 1
Catalog Number Mfr. No. ATCC® Format Price Quantity & Availability  
Catalog Number Mfr. No. ATCC® Format Price Quantity & Availability  
23-003-5007
View Documents
Microbiologics™
0317E3
ATCC 19404 Pellets Pack of 10 for $432.00
N/A
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
Epower
  • Lyophilized microorganism preparations
  • Biosafety Level: 1
  • Pellets can be used individually or combined for a mixed microorganism population challenge
  • Easily manipulated to deliver a wide variety of concentrations
  • Available in concentrations ranging from 102 to 108 CFU per pellet
  • ATCC™ 19404™†
Specifications
Specifications
10/Pk.
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. MicroBioLogics Inc. is licensed to use these trademarks and to sell products derived from ATCC cultures.