Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Microbiologics™ Epower™ Enterococcus faecalis ATCC™ 29212™†
For performing quality control functions including microbial detection, equipment calibration, method validation, and bioburden determination
$532.00
Specifications
Biosafety Level 1
ATCC® 29212
Format Lyophilized Pellet
For Use With (Application) For performing quality control functions including microbial detection, equipment calibration, method validation, and bioburden determination.
Quantity 10/Pk.
View More Specs
Products 1
Catalog Number Mfr. No. Mean Assay Value Price Quantity & Availability  
Catalog Number Mfr. No. Mean Assay Value Price Quantity & Availability  
12-111-251
View Documents
MicroBioLogicsSupplier Diversity Partner
0366E3CRM
1.0-9.9 + 3 CFU/pellet Each for $532.00
N/A
Description
Description
Epower
  • Lyophilized microorganism preparations
  • Biosafety Level: 1
  • Pellets can be used individually or combined for a mixed microorganism population challenge
  • Easily manipulated to deliver a wide variety of concentrations
  • Available in concentrations ranging from 102 to 108 CFU per pellet
  • ATCC™ 29212™†
Specifications
Specifications
1
Lyophilized Pellet
10/Pk.
29212
For performing quality control functions including microbial detection, equipment calibration, method validation, and bioburden determination.
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark and the ATCC catalog marks are trademarks of ATCC. Microbiologics, Inc. is licensed to use these trademarks and to sell products derived from ATCC cultures.