Learn More
Microbiologics™ Epower™ Enterococcus faecalis ATCC™ 29212™†
For performing quality control functions including microbial detection, equipment calibration, method validation, and bioburden determination
Manufacturer:  MicroBioLogicsSupplier Diversity Partner 0366E7CRM
 View more versions of this product
Catalog No. 12-111-252
Due to product restrictions, please to purchase or view availability for this product.
Disclaimers:
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark and the ATCC catalog marks are trademarks of ATCC. Microbiologics, Inc. is licensed to use these trademarks and to sell products derived from ATCC cultures.
Specifications Description & Specifications
Specifications
1
For performing quality control functions including microbial detection, equipment calibration, method validation, and bioburden determination.
1.0-9.9 + 7 CFU/pellet
29212
Lyophilized Pellet
10/Pk.