Microbiologics™ Epower™ Escherichia coli ATCC™ 43888™†
For performing quality control functions including microbial detection, equipment calibration, method validation, and bioburden determination
Manufacturer: MicroBioLogicsSupplier Diversity Partner 0795E3CRM
 View more versions of this product
Catalog No. 12-111-257
    N/A
Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 
Disclaimers:
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark and the ATCC catalog marks are trademarks of ATCC. Microbiologics, Inc. is licensed to use these trademarks and to sell products derived from ATCC cultures.
    Specifications
    Biosafety Level 2
    For Use With (Application) For performing quality control functions including microbial detection, equipment calibration, method validation, and bioburden determination.
    ATCC® 43888
    Format Lyophilized Pellet
    Mean Assay Value 1.0-9.9 + 3 CFU/pellet
    Quantity 10/Pk.
  • Description & Specifications
Related Products
Product Content Feedback