Learn More
Microbiologics™ Epower™ Escherichia coli Serotype O104:H4 ATCC™ BAA-2326™ (STEC)
Shop All Microbiologics Products

Description
- Biosafety level: 2
- Mean Assay Value: 1.0-9.93 CFU per pellet
Order Info
Available in concentrations ranging from 102 to 109 per pellet
Specifications
Specifications
| Biosafety Level | 2 |
| Format | Epower™ |
| For Use With (Application) | Food and Food Ingredients: Pathogen Testing |
| Quantity | 10 Pellets |
| Strain Designations | Serotype O104:H4 |
| Species | Escherichia coli |
†The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. Microbiologics™ is licensed to use these trademarks and to sell products derived from ATCC™ cultures
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.