Learn More
Microbiologics™ Epower Escherichia coli serotype O145:NM CDC 99-3311 (STEC)
Shop All Microbiologics Products

Specifications
Specifications
| Assay Range | 1.0-9.9E+03CFU/pellet |
| Biosafety Level | 2 |
| Quantity | 10/Pk. |
| Species | Escherichia coli |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.