Learn More
Microbiologics™ Epower™ Staphylococcus epidermidis, ATCC™ 12228™†
SDP
Lyophilized pellets add convenience to special projects and internal proficiency plans
Supplier:  Microbiologics™ 0371E4
 View more versions of this product
Catalog No. 23-003-5034
Due to product restrictions, please to purchase or view availability for this product.
Includes: Product warranty and peel-off certificate of assay
This item is not returnable. View return policy
Description
Description
Order Info
Available in vials containing 10 lyophilized pellets, with a range of concentration from 102 to 109 per pellet
Specifications
Specifications
Staphylococcus epidermidis
1
QC Microorganism
Product warranty and peel-off certificate of assay
Vial
12228
Lyophilized Pellet
Lyophilized pellets add convenience to special projects and internal proficiency plans.
1.0-9.9E+04 CFU per pellet
10/Pk.
Safety and Handling
Safety and Handling
ShelfLife : 10 to 20 Months
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. Microbiologics Inc. is licensed to use these trademarks and to sell products derived from ATCC™ cultures.