Learn More
Microbiologics™ Epower™ Kocuria rhizophila ATCC™ 9341™†
For performing quality control functions including microbial detection, equipment calibration, method validation, and bioburden determination
Specifications
Biosafety Level 1
ATCC® 9341
Format Lyophilized Pellet
For Use With (Application) For performing quality control functions including microbial detection, equipment calibration, method validation, and bioburden determination.
Quantity 10/Pk.
View More Specs
Products 2
Catalog Number Mfr. No. Mean Assay Value Price Quantity & Availability  
Catalog Number Mfr. No. Mean Assay Value Price Quantity & Availability  
12-111-264
View Documents
Microbiologics™
0688E3CRM
1.0-9.9 + 3 CFU/pellet This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Due to product restrictions, please to purchase or view availability for this product.
N/A
12-111-265
View Documents
Microbiologics™
0688E4CRM
1.0-9.9 + 4 CFU/pellet This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Due to product restrictions, please to purchase or view availability for this product.
N/A
Description
Description
Epower
  • Lyophilized microorganism preparations
  • Biosafety Level: 1
  • Pellets can be used individually or combined for a mixed microorganism population challenge
  • Easily manipulated to deliver a wide variety of concentrations
  • Available in concentrations ranging from 102 to 108 CFU per pellet
  • ATCC™ 9341™†
Specifications
Specifications
1
Lyophilized Pellet
10/Pk.
9341
For performing quality control functions including microbial detection, equipment calibration, method validation, and bioburden determination.
SDS
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark and the ATCC catalog marks are trademarks of ATCC. Microbiologics, Inc. is licensed to use these trademarks and to sell products derived from ATCC cultures.