Learn More
Microbiologics™ Epower™ Listeria monocytogenes ATCC™ 19115™†
SDP
For performing quality control functions including microbial detection, equipment calibration, method validation, and bioburden determination
Supplier:  Microbiologics™ 0687E3CRM
Catalog No. 12-111-272
Explore available promotions
Add to Cart
This item is not returnable. View return policy
Description
Description
Epower
  • Lyophilized microorganism preparations
  • Biosafety Level: 2
  • Pellets can be used individually or combined for a mixed microorganism population challenge
  • Easily manipulated to deliver a wide variety of concentrations
  • Available in concentrations ranging from 102 to 108 CFU per pellet
  • ATCC™ 19115™†
Specifications
Specifications
19115
Lyophilized Pellet
1.0-9.9 + 3 CFU/pellet
2
For performing quality control functions including microbial detection, equipment calibration, method validation, and bioburden determination.
10/Pk.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark and the ATCC catalog marks are trademarks of ATCC. Microbiologics, Inc. is licensed to use these trademarks and to sell products derived from ATCC cultures.