Learn More
Microbiologics™ Epower™ Penicillium aurantiogriseum ATCC™ 16025™†
For performing quality control functions including microbial detection, equipment calibration, method validation, and bioburden determination
Manufacturer:  Microbiologics™ 0794E3CRM
 View more versions of this product
Catalog No. 12-111-274
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
This item is not returnable. View return policy
Description
Description
Epower
  • Lyophilized microorganism preparations
  • Biosafety Level: 1
  • Pellets can be used individually or combined for a mixed microorganism population challenge
  • Easily manipulated to deliver a wide variety of concentrations
  • Available in concentrations ranging from 102 to 108 CFU per pellet
  • ATCC™ 16025™†
Specifications
Specifications
1
1.0-9.9 + 3 CFU/pellet
Penicillium aurantiogriseum
10/Pk.
16025
Penicillium aurantiogriseum
For performing quality control functions including microbial detection, equipment calibration, method validation, and bioburden determination.
Epower
SDS
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark and the ATCC catalog marks are trademarks of ATCC. Microbiologics, Inc. is licensed to use these trademarks and to sell products derived from ATCC cultures.