Learn More
Microbiologics™ Epower™ Salmonella enterica subsp. enterica serovar Abaetetuba ATCC™ 35640™†
For performing quality control functions including microbial detection, equipment calibration, method validation, and bioburden determination
Specifications
Biosafety Level 2
ATCC® 35640
For Use With (Application) For performing quality control functions including microbial detection, equipment calibration, method validation, and bioburden determination.
Species Salmonella enterica
View More Specs
Products 1
Catalog Number Mfr. No. Format Price Quantity & Availability  
Catalog Number Mfr. No. Format Price Quantity & Availability  
23-003-5005
View Documents
Microbiologics™
0817E2
Epower Pack of 10 for $421.50
N/A
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
Epower
  • Lyophilized microorganism preparations
  • Biosafety Level: 2
  • Pellets can be used individually or combined for a mixed microorganism population challenge
  • Easily manipulated to deliver a wide variety of concentrations
  • Available in concentrations ranging from 102 to 108 CFU per pellet
  • ATCC™ 35640™†
Specifications
Specifications
2
35640
For performing quality control functions including microbial detection, equipment calibration, method validation, and bioburden determination.
Salmonella enterica
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark and the ATCC catalog marks are trademarks of ATCC. Microbiologics, Inc. is licensed to use these trademarks and to sell products derived from ATCC cultures.