Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Microbiologics™ Epower™ Staphylococcus epidermidis ATCC™ 12228™†

For performing quality control functions including microbial detection, equipment calibration, method validation, and bioburden determination

Specifications

Biosafety Level 1
ATCC® 12228
Format Lyophilized Pellet
For Use With (Application) For performing quality control functions including microbial detection, equipment calibration, method validation, and bioburden determination.
Species Staphylococcus epidermidis
View More Specs

Products 1
Catalog Number Mfr. No. Mean Assay Value Price Quantity & Availability  
Catalog Number Mfr. No. Mean Assay Value Price Quantity & Availability  
12-111-293
View Documents
Microbiologics™
0371E3CRM
1.0-9.9 + 3 CFU/pellet Each for $543.50
N/A
Due to product restrictions, please to purchase or view availability for this product.
Description

Description

Epower

  • Lyophilized microorganism preparations
  • Biosafety Level: 1
  • Pellets can be used individually or combined for a mixed microorganism population challenge
  • Easily manipulated to deliver a wide variety of concentrations
  • Available in concentrations ranging from 102 to 108 CFU per pellet
  • ATCC™ 12228™†

Specifications

Specifications

1
Lyophilized Pellet
Staphylococcus epidermidis
12228
For performing quality control functions including microbial detection, equipment calibration, method validation, and bioburden determination.
10/Pk.

The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark and the ATCC catalog marks are trademarks of ATCC. Microbiologics, Inc. is licensed to use these trademarks and to sell products derived from ATCC cultures.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit