Learn More
Microbiologics™ EZ-CFU™ One-Step Microorganism Preparations
Lyophilized microorganism preparations deliver less than 100CFU per 0.1mL of hydrated suspension—no dilutions are required
Manufacturer:  Microbiologics™ 0318Z
 View more versions of this product
Catalog No. 23-003-828
Due to product restrictions, please to purchase or view availability for this product.
Specifications Description & Specifications
Specifications
13124
Product warranty and peel-off certificate of assay
EZ-CFU
EZ-CFU
Vial
Clostridium perfringens