Microbiologics™ EZ-FPC™ Microorganism Preparations

Lyophilized microorganism preparations deliver a predetermined assay value for qualitative or quantitative daily process controls

Manufacturer: Microbiologics™ 0306FPC

 View more versions of this product

Catalog No. 23-002-301

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Specifications

    For Use With (Application) Lyophilized microorganism preparations deliver a predetermined assay value for qualitative or quantitative daily process controls.
    ATCC® 13048
    Format EZ-FPC
    Includes Product warranty and peel-off certificate of assay
    Packaging Vial
    Product Type EZ-FPC
    Shelf Life 6 to 24 Months
    Species Enterobacter aerogenes

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit