Learn More
Microbiologics EZ-FPC Microorganism Preparations
Lyophilized microorganism preparations deliver a predetermined assay value for qualitative or quantitative daily process controls
Manufacturer: Microbiologics 0698FPC
Includes: Product warranty and peel-off Certificate of Assay
Description
- Available in 100 to 1000CFU per preparation (qualitative) or 1000 to 10,000CFU per preparation (quantitative)
- Daily process controls for food quality labs
- Traceable to reference cultures
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. MicroBioLogics Inc. is licensed to use these trademarks and to sell products derived from ATCC cultures.
Order Info
Packaged in a vial of a single assayed microorganism (10 lyophilized pellets in each vial), with an attached Certificate of Assay
Specifications
2601 | |
Saccharomyces cerevisiae | |
Lyophilized microorganism preparations deliver a predetermined assay value for qualitative or quantitative daily process controls. | |
EZ-FPC |
Product warranty and peel-off certificate of assay | |
EZ-FPC | |
6 to 24 Months | |
Vial |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.