Learn More
Microbiologics™ EZ-FPC™ Microorganism Preparations
Lyophilized microorganism preparations deliver a predetermined assay value for qualitative or quantitative daily process controls
Manufacturer:  Microbiologics™ 0813FPC
 View more versions of this product
Catalog No. 23-001-346
Due to product restrictions, please to purchase or view availability for this product.
Specifications Description & Specifications
Specifications
9338
EZ-FPC
Vial
Lactobacillus fermentum
Lyophilized microorganism preparations deliver a predetermined assay value for qualitative or quantitative daily process controls.
Product warranty and peel-off certificate of assay
EZ-FPC
Safety and Handling
ShelfLife : 6 to 24 Months