Learn More
Microbiologics™ Helix Elite™ Entamoeba histolytica Synthetic DNA
For use as positive control in molecular applications
Manufacturer:  Microbiologics™ HE0006S
Catalog No. 23-004-105
Add to cart
Includes: One vial of dried synthetic DNA or RNA (approximately 100 reactions), one vial of molecular standard water for rehydration and Certificate of Analysis
This item is not returnable. View return policy
Description
Description
  • Synthetic DNA/RNA molecular standard
  • 18D rRNA gene of Entamoeba histolytica
  • Dried DNA preserved with Biomatrica DNAstable™
Specifications
Specifications
Entamoeba histolytica Synthetic DNA