Learn More
Microbiologics™ Bacteroides ovatus ATCC™ 8483™†
SDP
For culture purposes and QC applications
Supplier:  Microbiologics™ 0400L
 View more versions of this product
Catalog No. 23-016-635
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
KWIK-STIK™
  • Clever design and accurate, repeatable results
  • Each device features a single strain of Bacteroides ovatus in a lyophilized pellet, a reservoir of hydrating fluid and inoculating swab
  • For added convenience, a peel-off identification label is included for easy documentation
  • Available in packs of 2 or 6
LYFO DISK™
  • The most economical reference stock culture option; lyophilized pellet containing a single strain of Bacteroides ovatus
  • Easy to use; simply re-hydrate the pellet and inoculate
  • Available in a re-sealable vial containing 6 pellets
  • ATCC™ 8483™†
Specifications
Specifications
Bacteroides ovatus
ATCC 8483
Lyfo-Disk QC Control
6/Pk.