Learn More
Microbiologics™ Escherichia coli NCIMB 8545
For culture purposes and QC applications
Manufacturer:  Microbiologics™ 0581P
 View more versions of this product
Catalog No. 23-013-517
Due to product restrictions, please to purchase or view availability for this product.
Specifications Description & Specifications
Specifications
2
CE, online certificate of analysis
Lyophilized Pellet
2/Pk.
2° to 8°C
NCIMB 8545
Culture purpose and QC application
KWIK-STIK
Escherichia coli
Safety and Handling
ShelfLife : 6 to 24 Months