Learn More
Microbiologics™ Kloeckera japonica ATCC™ 58370™†
SDP
For culture purposes and QC applications
Specifications
Species Kloeckera japonica
Product Type KWIK-STIK
View More Specs
Products 2
Catalog Number Mfr. No. ATCC® Product Type Price Quantity & Availability  
Catalog Number Mfr. No. ATCC® Product Type Price Quantity & Availability  
23-004-063
SDP
View Documents
Microbiologics™
01012P
58370 KWIK-STIK
Pack of 2 for $136.00
N/A
Due to product restrictions, please to purchase or view availability for this product.
23-016-007
SDP
View Documents
Microbiologics™
01012K
ATCC 58370 KWIK-STIK
Pack of 6 for $421.00
N/A
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
KWIK-STIK™
  • Clever design and accurate, repeatable results
  • Each device features a single Kloeckera japonica strain in a lyophilized pellet, a reservoir of hydrating fluid and inoculating swab
  • For added convenience, a peel-off identification label is included for easy documentation
  • Available in packs of 2
  • ATCC™ 58370™†
Specifications
Specifications
Kloeckera japonica
KWIK-STIK
Safety and Handling
Safety and Handling
ShelfLife : 6 to 24 Months