Learn More
Microbiologics™ Listeria innocua NCTC 11288
For culture purposes and QC applications
Manufacturer:  Microbiologics™ 0414P
Catalog No. 23-004-734
Due to product restrictions, please to purchase or view availability for this product.
This item is not returnable. View return policy
Description
Description
KWIK-STIK™
  • Clever design and accurate, repeatable results
  • Each device features a single Listeria innocua strain in a lyophilized pellet, a reservoir of hydrating fluid and inoculating swab
  • For added convenience, a peel-off identification label is included for easy documentation
  • Available in packs of 2
  • NCTC 11288
  • Serotype 6a
Specifications
Specifications
1
Listeria innocua
Listeria innocua QC microorganism
6 to 24 Months
Lyophilized Pellet
CE, online certificate of analysis
11288
2° to 8°C
Culture purpose and QC application
2U
Kwik-stik™