Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Microbiologics™ Listeria monocytogenes ATCC™ 19112™†
SDP

For culture purposes and QC applications

Supplier:  Microbiologics™ 0232P

 View more versions of this product

Catalog No. 23-003-203


Due to product restrictions, please to purchase or view availability for this product.

This item is not returnable. View return policy

Description

Description

KWIK-STIK™

  • Clever design and accurate, repeatable results
  • Each device features a single Listeria monocytogenes strain in a lyophilized pellet, a reservoir of hydrating fluid and inoculating swab
  • For added convenience, a peel-off identification label is included for easy documentation
  • Available in packs of 2
  • ATCC™ 19112™†

Specifications

Specifications

Listeria monocytogenes
2
2°C to 8°C
Kwik-stik™
Culture purpose and QC application
19112™*
CE, online certificate of analysis
Lyophilized Pellet
KWIK-STIK
2 U
Safety and Handling

Safety and Handling

ShelfLife : 6 to 24 Months

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit