Microbiologics™ Micrococcus luteus ATCC® 10240™†

For culture purposes and QC applications

Manufacturer: Microbiologics™ 0689X

 View all options for this product

Catalog No. 16-230-480


Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Biosafety Level 1
    ATCC® 10240
    Certifications/Compliance CE, certificate of assay
    For Use With (Application) Culture purpose and QC application
    Format Lyophilized Pellet
    Includes 2 Passages
    Media Type Kwik-stik™
    Product Type KWIK-STIK Plus
    Quantity 5/Pk.
    Shelf Life 6 to 24 Months
    Species Micrococcus luteus
    Storage Requirements 2° to 8°C


    • Clever design and accurate, repeatable results
    • Each device features a single Micrococcus luteus strain in a lyophilized pellet, a reservoir of hydrating fluid and inoculating swab
    • For added convenience, a peel-off identification label is included for easy documentation
    • Available in packs of 2 or 6, or 5 Plus pack
    • ATCC™ 10240™†

  • Description & Specifications

Related Products

Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit