Learn More
Microbiologics™ Micrococcus luteus NCIMB 8166
For culture purposes and QC applications
Manufacturer:  Microbiologics™ 0337P
 View more versions of this product
Catalog No. 23-004-785
Due to product restrictions, please to purchase or view availability for this product.
Specifications Description & Specifications
Specifications
1
CE, online certificate of analysis
Lyophilized Pellet
Micrococcus luteus QC microorganism
Micrococcus luteus
NCIMB 8166
Culture purpose and QC application
Kwik-stik™
2U
2° to 8°C
Safety and Handling
ShelfLife : 6 to 24 Months