Microbiologics™ Salmonella enterica subsp. enterica serovar Abony NCTC 6017

For culture purposes and QC applications

Manufacturer: Microbiologics™ 0890P

 View more versions of this product

Catalog No. 23-002-815

 N/A
    N/A

Part Not Found 


    Specifications

    Biosafety Level 2
    ATCC® 6017
    Certifications/Compliance CE, online certificate of analysis
    For Use With (Application) Culture purpose and QC application
    Format Lyophilized Pellet
    Media Type Kwik-stik™
    Product Type Salmonella enterica subspecies enterica serovar Abony QC microorganism
    Quantity 2U
    Shelf Life 6 to 24 Months
    Species Salmonella enterica
    Storage Requirements 2° to 8°C

    KWIK-STIK™

    • Clever design and accurate, repeatable results
    • Each device features a single Salmonella enterica subsp. enterica serovar Abony strain in a lyophilized pellet, a reservoir of hydrating fluid and inoculating swab
    • For added convenience, a peel-off identification label is included for easy documentation
    • Available in pack of 2
    LYFO DISK™
    • The most economical reference stock culture option; lyophilized pellet containing a single Salmonella enterica subsp. enterica serovar Abony strain
    • Easy to use; simply re-hydrate the pellet and inoculate
    • Available in a re-sealable vial containing 6 pellets
    • NCTC 6017

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit