Learn More
Microbiologics™ Staphylococcus aureus subsp. aureus ATCC® 33591™†
For culture purposes and QC applications
Manufacturer:  Microbiologics™ 0496X
 View more versions of this product
Catalog No. 23-017-157
Due to product restrictions, please to purchase or view availability for this product.
Specifications Description & Specifications
Specifications
2
CE, certificate of assay
Lyophilized Pellet
Kwik-stik™
5/Pk.
2° to 8°C
33591
Culture purpose and QC application
2 Passages
KWIK-STIK Plus
Staphylococcus aureus
Safety and Handling
ShelfLife : 6 to 24 Months