Learn More
Microbiologics™ Staphylococcus aureus subsp. aureus ATCC® 700699™†
For culture purposes and QC applications
Manufacturer:  Microbiologics™ 0158ROCHE
 View more versions of this product
Catalog No. 23-002-145
Due to product restrictions, please to purchase or view availability for this product.
Specifications Description & Specifications
Specifications
2
CE, online certificate of analysis
Lyophilized Pellet
KWIK-STIK, Two units at E4
Staphylococcus aureus
ATCC 700699
Culture purpose and QC application
Kwik-stik™
2/Pk.
2° to 8°C
Safety and Handling
ShelfLife : 6 to 24 Months