Learn More
Microbiologics™ Zygosaccharomyces bailii ATCC™ MYA-4549™†
SDP
For culture purposes and QC applications
$214.00
Specifications
Species Zygosaccharomyces bailii
Product Type KWIK-STIK
View More Specs
Products 2
Catalog Number Mfr. No. ATCC® Product Type Price Quantity & Availability  
Catalog Number Mfr. No. ATCC® Product Type Price Quantity & Availability  
23-004-078
SDP
View Documents
Microbiologics™
01011P
MYA-4549 KWIK-STIK
Pack of 2 for $214.00
Add to Cart
23-016-006
SDP
View Documents
Microbiologics™
01011K
ATCC MYA-4549 KWIK-STIK
Pack of 6 for $454.00
N/A
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
KWIK-STIK™
  • Clever design and accurate, repeatable results
  • Each device features a single Zygosaccharomyces bailii strain in a lyophilized pellet, a reservoir of hydrating fluid and inoculating swab
  • For added convenience, a peel-off identification label is included for easy documentation
  • Available in packs of 2 or 6
  • ATCC™ MYA-4549™†
Specifications
Specifications
Zygosaccharomyces bailii
KWIK-STIK
Safety and Handling
Safety and Handling
ShelfLife : 6 to 24 Months