Learn More
Microbiologics™ Klebsiella pneumoniae subsp. pneumoniae ATCC® 10031™†
For culture purposes and QC applications
Manufacturer:  Microbiologics™ 0684X
 View more versions of this product
Catalog No. 16-230-479
Due to product restrictions, please to purchase or view availability for this product.
Disclaimers:
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. MicroBioLogics Inc. is licensed to use these trademarks and to sell products derived from ATCC cultures.
Specifications Description & Specifications
Specifications
2
CE, certificate of assay
Lyophilized Pellet
Kwik-stik™
5/Pk.
2° to 8°C
10031
Culture purpose and QC application
2 Passages
KWIK-STIK
Klebsiella pneumoniae
Safety and Handling
ShelfLife : 6 to 24 Months