Learn More
Microbiologics™ Lab-Elite™ Listeria welshimeri ATCC™ 35897™†
SDP
For validation and verification of processes and instruments
Supplier:  Microbiologics™ 0816CRM
Catalog No. 23-002-068
Due to product restrictions, please to purchase or view availability for this product.
This item is not returnable. View return policy
Description
Description
Lab Elite™
  • Delivered in an easy-to-use KWIK-STIK™ format
  • Traceable to reference culture
  • Highly characterized Listeria welshimeri
  • For use in ISO 17025 accredited labs and supported by the highest accreditations in the industry
  • ATCC™ 35897™†
Specifications
Specifications
Listeria welshimeri
1
For validation and verification of processes and instruments.
Single
35897™*
KWIK-STIK
Lyophilized microbe pellet, reservoir of hydrating fluid, inoculating swab (KWIK-STIK format)
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark and the ATCC catalog marks are trademarks of ATCC. Microbiologics, Inc. is licensed to use these trademarks and to sell products derived from ATCC cultures.