Learn More
Microbiologics™ Lab-Elite™ Staphylococcus epidermidis ATCC™ 14990™†
For validation and verification of processes and instruments
Manufacturer:  Microbiologics™ 0412CRM
Catalog No. 23-002-075
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
Lab Elite™
  • Delivered in an easy-to-use KWIK-STIK™ format
  • Traceable to reference culture
  • Highly characterized Staphylococcus epidermidis
  • For use in ISO 17025 accredited labs and supported by the highest accreditations in the industry
  • ATCC™ 14990™†
Specifications
Specifications
1
Lyophilized microbe pellet, reservoir of hydrating fluid, inoculating swab (KWIK-STIK format)
For validation and verification of processes and instruments.
Single
14990™*
Staphylococcus epidermidis
KWIK-STIK
SDS
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark and the ATCC catalog marks are trademarks of ATCC. Microbiologics, Inc. is licensed to use these trademarks and to sell products derived from ATCC cultures.