Learn More
Listeria ivanovii subsp. ivanovii ATCC 19119, Microbiologics
For culture purposes and QC applications
Manufacturer: MicroBioLogics 0815P
Description
KWIK-STIK
- Clever design and accurate, repeatable results
- Each device features a single strain of Listeria ivanovii subsp. ivanovii in a lyophilized pellet, a reservoir of hydrating fluid and inoculating swab
- For added convenience, a peel-off identification label is included for easy documentation
- Available in packs of 2
- The most economical reference stock culture option; lyophilized pellet containing a single strain of Listeria ivanovii subsp. ivanovii
- Easy to use; simply re-hydrate the pellet and inoculate
- Available in a re-sealable vial containing 6 or 10 pellets
- ATCC 19119
Specifications
2 | |
Listeria ivanovii | |
Listeria ivanovii subspecies ivanovii QC microorganism | |
6 to 24 Months | |
Lyophilized Pellet | |
CE, online certificate of analysis |
19119 | |
2° to 8°C | |
Culture purpose and QC application | |
2U | |
Kwik-stik™ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. MicroBioLogics Inc. is licensed to use these trademarks and to sell products derived from ATCC cultures.