Learn More
Listeria ivanovii subsp. ivanovii ATCC™ 19119™†, Microbiologics™
SDP
For culture purposes and QC applications
Supplier:  MicroBioLogics 0815L
 View more versions of this product
Catalog No. 23-016-835
Due to product restrictions, please to purchase or view availability for this product.
This item is not returnable. View return policy
Description
Description
KWIK-STIK™
  • Clever design and accurate, repeatable results
  • Each device features a single strain of Listeria ivanovii subsp. ivanovii in a lyophilized pellet, a reservoir of hydrating fluid and inoculating swab
  • For added convenience, a peel-off identification label is included for easy documentation
  • Available in packs of 2
LYFO DISK™
  • The most economical reference stock culture option; lyophilized pellet containing a single strain of Listeria ivanovii subsp. ivanovii
  • Easy to use; simply re-hydrate the pellet and inoculate
  • Available in a re-sealable vial containing 6 or 10 pellets
  • ATCC™ 19119™†
Specifications
Specifications
19119
CE, online certificate of analysis
Lyophilized Pellet
QC Microorganism
6 Pellets
2
2°C to 8°C
Lyfo disk™
Culture purpose and QC application
Safety and Handling
Safety and Handling
ShelfLife : 6 to 24 Months
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. MicroBioLogics Inc. is licensed to use these trademarks and to sell products derived from ATCC cultures.